siRNA Details
Gene Symbol: RAMP2-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_024461.1 | 2 | 611 | |||
NR_024462.1 | 3 | 490 | ||||
GenBank mRNA | AK057121.1 | - | 1 | 639 | ||
BC001607.1 | - | 2 | 457 | |||
BC018638.2 | - | 3 | 490 | |||
BC033118.1 | - | 3 | 490 | |||
BC034924.1 | - | 2 | 365 | |||
BC041021.1 | - | 2 | 458 |
