siRNA Details
Gene Symbol: SNHG12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_024127.1 | 6 | 660 | |||
GenBank mRNA | AF277181.1 | - | 6 | 448 | ||
AK092096.1 | - | 3 | 1829 | |||
BC022205.1 | - | 6 | 1289 | |||
BC034611.1 | - | 4 | 1511 | |||
BC045792.1 | - | 5 | 1019 | |||
BC064589.1 | - | 4 | 1550 | |||
BC068048.1 | - | 6 | 660 | |||
BC107730.1 | - | 3 | 1052 | |||
KJ782215.1 | - | 7 | 1789 |
