siRNA Details
Gene Symbol: LOC107984014 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_017012898.1 | XP_016868387.1 | 5 | 512 |
Gene Symbol: LOC101929998 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK127966.1 | - | 1 | 216 |
