siRNA Details
Gene Symbol: PDCD4-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_026932.1 | 1 | 602 | |||
GenBank mRNA | AK093606.1 | - | 1 | 602 | ||
AK097703.1 | - | 1 | 1477 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: PDCD4-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_026932.1 | 1 | 602 | |||
GenBank mRNA | AK093606.1 | - | 1 | 602 | ||
AK097703.1 | - | 1 | 1477 |