siRNA Details
Gene Symbol: SERTAD4-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_024337.1 | 2 | 627 | |||
GenBank mRNA | BC066352.1 | - | 2 | 626 | ||
BC094794.1 | - | 1 | 2527 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: SERTAD4-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_024337.1 | 2 | 627 | |||
GenBank mRNA | BC066352.1 | - | 2 | 626 | ||
BC094794.1 | - | 1 | 2527 |