siRNA Details
Gene Symbol: SPATA42 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_034168.1 | 2 | 384 | |||
NR_049777.1 | 2 | 384 | ||||
GenBank mRNA | AA401510.1 | - | 2 | 357 | ||
AY312144.1 | - | 2 | 384 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: SPATA42 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_034168.1 | 2 | 384 | |||
NR_049777.1 | 2 | 384 | ||||
GenBank mRNA | AA401510.1 | - | 2 | 357 | ||
AY312144.1 | - | 2 | 384 |