siRNA Details
Gene Symbol: LINC00963 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_038955.1 | 1 | 555 | |||
GenBank mRNA | AK024177.1 | - | 1 | 108 | ||
AK092686.1 | - | 1 | 13 | |||
BC044229.1 | - | 1 | 115 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | AU122149.1 | - | 1 | 108 | Chr. 9 - 129489107 - 129502906 [+] on Build GRCh38 |
