siRNA Details
Gene Symbol: PSMB8-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_037173.1 | 3 | 473 | |||
NR_037174.1 | 2 | 335 | ||||
NR_037175.1 | 3 | 308 | ||||
NR_037176.1 | 2 | 247 | ||||
GenBank mRNA | BI906764.1 | - | 2 | 200 | ||
BU539272.1 | - | 2 | 283 | |||
CK904130.1 | - | 3 | 426 | |||
CR977230.1 | - | 3 | 263 |
