siRNA Details
Gene Symbol: TNRC6C-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_040071.1 | 1 | 1681 | |||
GenBank mRNA | AK092916.1 | - | 1 | 534 | ||
AK094898.1 | - | 1 | 1153 | |||
AK123771.1 | - | 1 | 1859 | |||
AY129024.1 | - | 1 | 217 | |||
BC041417.2 | - | 1 | 1682 |
