siRNA Details
Gene Symbol: TNRC6C-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_040071.1 | 2 | 2954 | |||
GenBank mRNA | AK092916.1 | - | 2 | 2027 | ||
AK094898.1 | - | 2 | 2426 | |||
AK127699.1 | - | 1 | 2205 | |||
AY129024.1 | - | 1 | 2714 | |||
BC041417.2 | - | 2 | 2955 |
