siRNA Details
Gene Symbol: FLJ45079 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_028337.1 | 2 | 3420 | |||
GenBank mRNA | AK127023.1 | - | 2 | 3420 | ||
AK128332.1 | - | 1 | 2293 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: FLJ45079 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_028337.1 | 2 | 3420 | |||
GenBank mRNA | AK127023.1 | - | 2 | 3420 | ||
AK128332.1 | - | 1 | 2293 |