siRNA Details
Gene Symbol: C9orf170 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001001709.2 | NP_001001709.1 | 2 | 1094 | ||
GenBank mRNA | AK127445.1 | - | 2 | 1094 | ||
BC132741.1 | - | 2 | 1083 | 40146763 | ||
BC136791.1 | - | 2 | 1083 | 9020781 |
