siRNA Details
Gene Symbol: C9orf170 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001001709.2 | NP_001001709.1 | 2 | 1784 | ||
GenBank mRNA | AK127445.1 | - | 2 | 1784 | ||
BC136791.1 | - | 2 | 1773 | 9020781 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: C9orf170 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001001709.2 | NP_001001709.1 | 2 | 1784 | ||
GenBank mRNA | AK127445.1 | - | 2 | 1784 | ||
BC136791.1 | - | 2 | 1773 | 9020781 |