siRNA Details
Gene Symbol: LINC01123 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_046110.1 | 2 | 781 | |||
NR_046111.1 | 2 | 781 | ||||
GenBank mRNA | AK056084.1 | - | 2 | 698 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: LINC01123 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_046110.1 | 2 | 781 | |||
NR_046111.1 | 2 | 781 | ||||
GenBank mRNA | AK056084.1 | - | 2 | 698 |