siRNA Details
Gene Symbol: MAMDC2-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_040076.1 | 4 | 638 | |||
GenBank mRNA | BC043510.1 | - | 4 | 582 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BI830228.1 | - | 4 | 581 | Chr. 9 - 70153929 - 70175832 [-] on Build GRCh38 |
