siRNA Details
Gene Symbol: CHKB-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_021492.2 | 2 | 536 | |||
GenBank mRNA | BE393070.1 | - | 2 | 438 | ||
BJ989458.1 | - | 2 | 417 | |||
CR980518.1 | - | 2 | 440 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | HY124358.1 | - | 2 | 439 | Chr. 22 - 50583124 - 50583885 [+] on Build GRCh38 |
