siRNA Details
Gene Symbol: DPP9-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001242901.1 | NP_001229830.1 | 3 | 2870 | ||
GenBank mRNA | AK126095.1 | - | 3 | 2870 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: DPP9-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001242901.1 | NP_001229830.1 | 3 | 2870 | ||
GenBank mRNA | AK126095.1 | - | 3 | 2870 |