siRNA Details
Gene Symbol: EWSAT1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_026949.1 | 3 | 1235 | |||
GenBank mRNA | AK091087.1 | - | 3 | 1223 | ||
AK097560.1 | - | 4 | 1064 | |||
AK292174.1 | - | 3 | 788 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: EWSAT1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_026949.1 | 3 | 1235 | |||
GenBank mRNA | AK091087.1 | - | 3 | 1223 | ||
AK097560.1 | - | 4 | 1064 | |||
AK292174.1 | - | 3 | 788 |