siRNA Details
Gene Symbol: C1orf220 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_033186.1 | 3 | 1893 | |||
GenBank mRNA | AK092849.1 | - | 3 | 1893 | ||
BC136727.1 | - | 3 | 1874 | 9020717 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: C1orf220 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_033186.1 | 3 | 1893 | |||
GenBank mRNA | AK092849.1 | - | 3 | 1893 | ||
BC136727.1 | - | 3 | 1874 | 9020717 |