siRNA Details
Gene Symbol: LINC00671 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_027254.1 | 4 | 546 | |||
GenBank mRNA | AK055784.1 | - | 4 | 656 | ||
AK298293.1 | - | 3 | 477 | |||
BC122867.1 | - | 3 | 376 | |||
BC122868.1 | - | 4 | 468 | |||
DC361857.1 | - | 3 | 477 |
