siRNA Details
Gene Symbol: LIPE-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_073180.1 | 2 | 89 | |||
GenBank mRNA | AK311181.1 | - | 2 | 63 | ||
BC037865.2 | - | 2 | 948 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | HY353777.1 | - | 1 | 83 | Chr. 9 - 72355928 - 72360168 [+] on Build GRCh38 |
