siRNA Details
Gene Symbol: VAC14-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_034083.2 | 6 | 1742 | |||
GenBank mRNA | AK095020.1 | - | 6 | 1668 | ||
BC035152.1 | - | 5 | 4370 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: VAC14-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_034083.2 | 6 | 1742 | |||
GenBank mRNA | AK095020.1 | - | 6 | 1668 | ||
BC035152.1 | - | 5 | 4370 |