siRNA Details
Gene Symbol: KANSL1-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_034172.1 | 2 | 238 | |||
GenBank mRNA | AF088058.1 | - | 1 | 382 | ||
BC020847.1 | - | 2 | 238 | |||
BC066350.1 | - | 2 | 235 | |||
FJ009034.1 | - | 2 | 222 |
