siRNA Details
Gene Symbol: LOC152225 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_026934.1 | 1 | 48 | |||
GenBank mRNA | AK090588.1 | - | 1 | 48 | ||
BG249468.1 | - | 1 | 49 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: LOC152225 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_026934.1 | 1 | 48 | |||
GenBank mRNA | AK090588.1 | - | 1 | 48 | ||
BG249468.1 | - | 1 | 49 |