siRNA Details
Gene Symbol: LINC01588 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_126496.1 | 4 | 4043 | |||
NR_126497.1 | 3 | 1440 | ||||
NR_126498.1 | 3 | 2067 | ||||
NR_126499.1 | 3 | 272 | ||||
GenBank mRNA | AK090420.1 | - | 4 | 4043 | ||
AK091232.1 | - | 3 | 2066 | |||
BC128083.1 | - | 4 | 2028 | 40075904 | ||
BC144351.1 | - | 3 | 1439 | 9052871 |
