siRNA Details
Gene Symbol: CD27-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_015382.2 | 6 | 1957 | |||
GenBank mRNA | AK056500.1 | - | 6 | 1957 | ||
AK128574.1 | - | 2 | 2829 | |||
BC033973.1 | - | 7 | 930 | |||
BC041960.1 | - | 3 | 1253 |
