siRNA Details
Gene Symbol: DHRS4-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_023921.2 | 2 | 1078 | |||
NR_023922.2 | 2 | 819 | ||||
NR_023923.2 | 3 | 787 | ||||
NR_023924.2 | 2 | 878 | ||||
GenBank mRNA | BC080582.1 | - | 2 | 805 | ||
BX247982.1 | - | 2 | 548 | |||
BX447332.2 | - | 2 | 1003 | |||
BX647964.1 | - | 3 | 755 | |||
HQ845197.1 | - | 3 | 773 | |||
HQ845198.1 | - | 3 | 773 | |||
HQ845199.1 | - | 2 | 858 | |||
JQ979294.1 | - | 3 | 773 | |||
KJ545484.1 | - | 2 | 714 | |||
KJ574270.1 | - | 2 | 505 | |||
KJ574271.1 | - | 2 | 638 | |||
KJ574272.1 | - | 2 | 637 | |||
KJ574273.1 | - | 2 | 797 | |||
KJ574274.1 | - | 2 | 841 | |||
KJ599455.1 | - | 2 | 571 | |||
KJ599456.1 | - | 2 | 828 | |||
KJ599457.1 | - | 2 | 530 | |||
KJ599458.1 | - | 1 | 324 | |||
KJ599459.1 | - | 3 | 746 | |||
KJ599460.1 | - | 2 | 872 | |||
KJ599461.1 | - | 3 | 756 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BX461600.2 | - | 2 | 549 | Chr. 14 - 23940338 - 23953668 [-] on Build GRCh38 |
