siRNA Details
Gene Symbol: DHRS4-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_023921.2 | 2 | 1115 | |||
NR_023922.2 | 2 | 856 | ||||
NR_023923.2 | 3 | 824 | ||||
NR_023924.2 | 2 | 915 | ||||
GenBank mRNA | BC080582.1 | - | 2 | 842 | ||
BX247982.1 | - | 2 | 585 | |||
BX447332.2 | - | 2 | 1040 | |||
BX647964.1 | - | 3 | 792 | |||
HQ845197.1 | - | 3 | 810 | |||
HQ845198.1 | - | 3 | 810 | |||
HQ845199.1 | - | 2 | 895 | |||
JQ979294.1 | - | 3 | 810 | |||
KJ545484.1 | - | 2 | 751 | |||
KJ574270.1 | - | 2 | 542 | |||
KJ574271.1 | - | 2 | 675 | |||
KJ574272.1 | - | 2 | 674 | |||
KJ574273.1 | - | 2 | 834 | |||
KJ574274.1 | - | 2 | 878 | |||
KJ599455.1 | - | 2 | 608 | |||
KJ599456.1 | - | 2 | 865 | |||
KJ599457.1 | - | 2 | 567 | |||
KJ599458.1 | - | 1 | 361 | |||
KJ599459.1 | - | 3 | 783 | |||
KJ599460.1 | - | 2 | 909 | |||
KJ599461.1 | - | 3 | 793 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BX461600.2 | - | 2 | 586 | Chr. 14 - 23940338 - 23953668 [-] on Build GRCh38 |
