siRNA Details
Gene Symbol: LINC01139 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_015407.1 | 2 | 731 | |||
GenBank mRNA | BC044843.1 | - | 2 | 731 | ||
BC064529.1 | - | 2 | 737 | |||
BC073894.1 | - | 2 | 708 | |||
BC113688.1 | - | 1 | 447 | |||
BC113690.1 | - | 1 | 447 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BI761548.1 | - | 2 | 729 | Chr. 1 - 238481108 - 238486017 [-] on Build GRCh38 |
