siRNA Details
Gene Symbol: KIRREL3-AS3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_040078.1 | 3 | 1751 | |||
GenBank mRNA | AK097543.1 | - | 3 | 1752 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: KIRREL3-AS3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_040078.1 | 3 | 1751 | |||
GenBank mRNA | AK097543.1 | - | 3 | 1752 |