siRNA Details
Gene Symbol: KIRREL3-AS3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_040078.1 | 1 | 52 | |||
GenBank mRNA | AK097543.1 | - | 1 | 52 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | DB036346.1 | - | 1 | 52 | Chr. 11 - 127002910 - 127003436 [+] on Build GRCh38 |
