siRNA Details
Gene Symbol: LOC101928254 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_110182.1 | 4 | 618 | |||
GenBank mRNA | EF565094.1 | - | 4 | 619 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: LOC101928254 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_110182.1 | 4 | 618 | |||
GenBank mRNA | EF565094.1 | - | 4 | 619 |