siRNA Details
Gene Symbol: LOC101927285 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_110219.1 | 3 | 264 | |||
GenBank mRNA | AK056269.1 | - | 3 | 264 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | DA703187.1 | - | 3 | 264 | Chr. 2 - 59217708 - 59277836 [+] on Build GRCh38 |
