siRNA Details
Gene Symbol: CLDN10-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_046533.1 | 4 | 666 | |||
GenBank mRNA | BC024972.1 | - | 4 | 667 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: CLDN10-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_046533.1 | 4 | 666 | |||
GenBank mRNA | BC024972.1 | - | 4 | 667 |