siRNA Details
Gene Symbol: DIP2A-IT1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_046400.1 | 1, 2 | 54 | |||
GenBank mRNA | BM929894.1 | - | 1, 2 | 54 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: DIP2A-IT1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_046400.1 | 1, 2 | 54 | |||
GenBank mRNA | BM929894.1 | - | 1, 2 | 54 |