siRNA Details
Gene Symbol: CCDC18-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_034089.1 | 4, 5 | 358 | |||
GenBank mRNA | AL832786.1 | - | 4, 5 | 358 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | DA948596.1 | - | 4, 5 | 522 | Chr. 1 - 93339236 - 93345790 [-] on Build GRCh38 |
