siRNA Details
Gene Symbol: IL10RB-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK123722.1 | - | 2 | 935 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | CB046228.1 | - | 1 | 523 | Chr. 21 - 33265147 - 33265664 [-] on Build GRCh38 |
