siRNA Details
Gene Symbol: IL10RB-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK123722.1 | - | 2 | 1491 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | AI004240.1 | - | 2 | 377 | Chr. 21 - 33264534 - 33265559 [-] on Build GRCh38 |
