siRNA Details
Gene Symbol: ZNF32-AS3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_038867.1 | 2 | 357 | |||
GenBank mRNA | BC033403.1 | - | 2 | 357 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: ZNF32-AS3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_038867.1 | 2 | 357 | |||
GenBank mRNA | BC033403.1 | - | 2 | 357 |