siRNA Details
Gene Symbol: MIR181A2HG | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_038975.1 | 2 | 336 | |||
GenBank mRNA | AB593023.1 | - | 2 | 338 | ||
BM716898.1 | - | 2 | 378 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | EL947414.1 | - | 2 | 362 | Chr. 9 - 124658459 - 124698616 [+] on Build GRCh38 |
