siRNA Details
Gene Symbol: MIR181A2HG | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_038975.1 | 1 | 297 | |||
GenBank mRNA | AB593023.1 | - | 1 | 299 | ||
BM716898.1 | - | 1 | 266 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | EL947414.1 | - | 1 | 323 | Chr. 9 - 124658459 - 124698616 [+] on Build GRCh38 |
