siRNA Details
Gene Symbol: INHBA-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_027119.1 | 3 | 714 | |||
GenBank mRNA | AK096266.1 | - | 1 | 1609 | ||
BC047446.1 | - | 3 | 711 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BI602851.1 | - | 3 | 715 | Chr. 7 - 41693952 - 41712062 [+] on Build GRCh38 |
