siRNA Details
Gene Symbol: MIR3681HG | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_110197.1 | 3 | 277 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BX090984.1 | - | 2 | 201 | Chr. 2 - 12106582 - 12131548 [+] on Build GRCh38 |
