siRNA Details
Gene Symbol: C5orf66 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001277348.1 | NP_001264277.1 | 6 | 2137 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: C5orf66 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001277348.1 | NP_001264277.1 | 6 | 2137 |