siRNA Details
Gene Symbol: LOC105374266 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_135537.1 | 2 | 171 | |||
NR_135538.1 | 2 | 171 | ||||
GenBank mRNA | BX364319.2 | - | 2 | 171 | ||
BX403058.2 | - | 2 | 171 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: LOC105374266 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_135537.1 | 2 | 171 | |||
NR_135538.1 | 2 | 171 | ||||
GenBank mRNA | BX364319.2 | - | 2 | 171 | ||
BX403058.2 | - | 2 | 171 |