siRNA Details
Gene Symbol: LOC100507654 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BC034289.1 | - | 3 | 188 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BG721018.1 | - | 3 | 193 | Chr. 13 - 100537323 - 100580113 [-] on Build GRCh38 |
