siRNA Details
Gene Symbol: LINC01272 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001278655.1 | NP_001265584.1 | 4 | 1537 | ||
XM_006723680.3 | XP_006723743.1 | 4 | 1550 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: LINC01272 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001278655.1 | NP_001265584.1 | 4 | 1537 | ||
XM_006723680.3 | XP_006723743.1 | 4 | 1550 |