siRNA Details
Gene Symbol: LINC00620 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_027103.1 | 6 | 662 | |||
GenBank mRNA | BC039529.1 | - | 6 | 662 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BM564239.1 | - | 6 | 681 | Chr. 3 - 13650721 - 13746532 [+] on Build GRCh38 |
