siRNA Details
Gene Symbol: LINC01015 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_037179.1 | 3 | 1692 | |||
NR_037180.1 | 2 | 1597 | ||||
NR_037181.1 | 4 | 781 | ||||
GenBank mRNA | AK097713.1 | - | 3 | 1678 | ||
AK126097.1 | - | 1 | 3027 |
