siRNA Details
Gene Symbol: LINC01015 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_037179.1 | 2 | 218 | |||
NR_037181.1 | 2 | 218 | ||||
GenBank mRNA | AK097713.1 | - | 2 | 204 | ||
AK126097.1 | - | 1 | 545 | |||
CD358111.1 | - | 2 | 148 |
